Acetamide ≥99%

Supplier: BeanTown Chemical

144855-1KG 144855-250G 144855-5KG
BT144855-250GEA 38.82 USD
BT144855-250G BT144855-5KG BT144855-1KG
Acetamide ≥99%
Acetamide

  • RTECS: AB4025000
  • Crystalline

Formula: C₂H₅NO
MW: 59.07 g/mol
Boiling Pt: 221 °C
Melting Pt: 78…80 °C
MDL Number: MFCD00008023
CAS Number: 60-35-5
EINECS: 200-473-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR