Platinum disulfide
Supplier: BeanTown Chemical
Synonyms:
Platinum(IV) sulphide
, Platinum (IV) sulfide
, Platinum(IV) sulfide
, Platinum (IV) sulphide
143940-1G
143940-5G
BT143940-5GEA
1398.12
USD
BT143940-5G
BT143940-1G
Platinum disulfide
Platinum disulphide
- Powder
Formula:
PtS₂ MW: 259.21 g/mol Melting Pt: 225…250 °C (dec.) |
MDL Number:
MFCD00016293 CAS Number: 12038-21-0 EINECS: 234-876-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...