Isoluminol (4-aminophthalhydrazide) ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Amino-2,3-dihydro-1,4-phthalazinedione
, 4-Aminophthaloylhydrazine
, Isoluminol
, 4-Aminophthalhydrazide
143630-1G
143630-5G
BT143630-1GEA
113.94
USD
BT143630-1G
BT143630-5G
Isoluminol (4-aminophthalhydrazide) ≥98%
Isoluminol (4-Aminophthalhydrazide)
- Air Sensitive
- Powder
Formula:
C₈H₇N₃O₂ MW: 177.16 Melting Pt: 300 °C |
MDL Number:
MFCD00010560 CAS Number: 3682-14-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...