2-Ethylimidazole ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Ethyl-1H-imidazole
143620-500G
143620-100G
BT143620-100GEA
49.8
USD
BT143620-100G
BT143620-500G
2-Ethylimidazole ≥98%
2-Ethylimidazole
- Flakes
Formula:
C₅H₈N₂ MW: 96.13 g/mol Boiling Pt: 268 °C Melting Pt: 78…81 °C Flash Pt: 159 °C (318 °F) |
MDL Number:
MFCD00005192 CAS Number: 1072-62-4 EINECS: 214-011-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...