Potassium niobate ≥99.999% (trace metals basis)

143605-25G 143605-100G
BT143605-25GEA 131.81 USD
BT143605-25G BT143605-100G
Potassium niobate ≥99.999% (trace metals basis)
Potassium niobate

  • RTECS: TT3500000
  • Powder

Formula: KNbO₃
MDL Number: MFCD00049651
CAS Number: 12030-85-2
EINECS: 234-744-4

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR