Cobalt aluminate

Supplier: BeanTown Chemical

143335-500G 143335-100G
BT143335-500GEA 123.6 USD
BT143335-500G BT143335-100G
Cobalt aluminate
Cobalt aluminate

  • -200 Mesh Powder

Formula: Al₂Co₂O₅
MDL Number: MFCD00016016
CAS Number: 12672-27-4
EINECS: 235-762-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR