2-Adamantanol ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Tricyclo[3.3.1.1(3,7)]decanol
, Tricyclo[3.3.1.1(3,7)]decan-2-ol
, 2-Hydroxytricyclo[3.3.1.1(3,7)]decane
143320-25G
143320-5G
BT143320-25GEA
221.24
USD
BT143320-25G
BT143320-5G
2-Adamantanol ≥98%
2-Adamantanol
- Crystalline/Powder
Formula:
C₁₀H₁₆O MW: 152.24 g/mol Melting Pt: 258…262 °C |
MDL Number:
MFCD00074744 CAS Number: 700-57-2 EINECS: 211-846-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...