Barium carbonate ≥99.5%

Supplier: BeanTown Chemical

142975-1KG 142975-50G 142975-250G
BT142975-250GEA 46.43 USD
BT142975-250G BT142975-50G BT142975-1KG
Barium carbonate ≥99.5%
Barium carbonate

  • RTECS: CQ8600000
  • 1 Micron Powder

Formula: BaCO₃
MW: 197.34 g/mol
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR