Iridium ≥99.5% (trace metals basis), wire Ø 1.0 mm

142960-1CM 142960-25CM 142960-5CM
BT142960-5CMEA 651.99 USD
BT142960-5CM BT142960-25CM BT142960-1CM
Iridium ≥99.5% (trace metals basis), wire Ø 1.0 mm
Iridium

  • Solid

Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 4130 °C
Melting Pt: 2450 °C
Density: 22.65 g/mL
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR