2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, Technical Grade
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Heptafluorobutyraldehyde hydrate
, 2,2,3,3,4,4,4-Heptafluorobutanal hydrate
142770-5G
142770-25G
142770-1G
BT142770-1GEA
43.84
USD
BT142770-1G
BT142770-25G
BT142770-5G
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, Technical Grade
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate
Formula:
CF₃CF₂CF₂CHO·xH₂O MW: 198.04 g/mol (anhy) |
MDL Number:
MFCD00150588 CAS Number: 375-02-0 EINECS: 206-783-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...