(R)-(+)-2-Hydroxy-1,2,2-triphenylethyl acetate ≥97%

Supplier: BeanTown Chemical

Synonyms: (R)-(+)-1,1,2-Triphenyl-1,2-ethanediol 2-acetate

142685-5G
BT142685-5GEA 236.38 USD
BT142685-5G
(R)-(+)-2-Hydroxy-1,2,2-triphenylethyl acetate ≥97%
(R)-(+)-2-Hydroxy-1,2,2-triphenylethyl acetate

  • Optical rotation: [α]20/D +212°, c = 1 in pyridine
  • Solid

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR