Zinc(II) dibutyldithiocarbamate ≥97%

Supplier: BeanTown Chemical

Synonyms: Dibutyldithiocarbamic Acid Zinc(II) Salt

142660-25G
BT142660-25GEA 36.46 USD
BT142660-25G
Zinc(II) dibutyldithiocarbamate ≥97%
Zinc(II) dibutyldithiocarbamate

  • RTECS: ZH0175000
  • Solid

Formula: C₉H₁₉NS₂
MW: 474.12 g/mol
Melting Pt: 104…110 °C
MDL Number: MFCD00067274
CAS Number: 136-23-2
EINECS: 205-232-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR