6-Amino-1,3-dimethyluracil ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Amino-1,3-Dimethyluracil

142370-100G
BT142370-100GEA 74.01 USD
BT142370-100G
6-Amino-1,3-dimethyluracil ≥98%
6-Amino-1,3-dimethyluracil

CAS: 6642-31-5; EC No: 229-662-0; MDL No: MFCD00006552; RTECS: YQ8755000
Powder; Molecular Formula: C6H9N3O2; MW: 155.15
Melting Point: 295° (decomposes)

Formula: C₆H₉N₃O₂
Melting Pt: 295 °C (dec.)
MDL Number: MFCD00006552
CAS Number: 6642-31-5
EINECS: 229-662-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR