1,3-Di-4-piperidylpropane ≥97%

Supplier: BeanTown Chemical

Synonyms: 4,4'-Trimethylenedipiperidine1,3-Bis(4-piperidyl)propane

142220-25G 142220-100G
BT142220-25GEA 37.99 USD
BT142220-25G BT142220-100G
1,3-Di-4-piperidylpropane ≥97%
1,3-Di-4-piperidylpropane

  • Hygroscopic
  • Powder

Formula: C₁₃H₂₆N₂
MW: 210.37 g/mol
Melting Pt: 63…68 °C
MDL Number: MFCD00038013
CAS Number: 16898-52-5
EINECS: 240-941-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR