trans-4-(4-Ethylcyclohexyl)benzonitrile ≥99%

Supplier: BeanTown Chemical

142175-5G 142175-1G
BT142175-5GEA 304.67 USD
BT142175-5G BT142175-1G
trans-4-(4-Ethylcyclohexyl)benzonitrile ≥99%
trans-4-(4-Ethylcyclohexyl)benzonitrile

  • Solid

Formula: C₁₅H₁₉N
Melting Pt: 41 °C
MDL Number: MFCD09750932
CAS Number: 72928-54-2
EINECS: 277-084-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR