Bis(triphenylphosphine)nickel(II) chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Bis(triphenylphosphine)nickel(II)chloride
, Dichlorobis(triphenylphosphine)nickel(II)
141785-50G
141785-10G
BT141785-50GEA
149.56
USD
BT141785-50G
BT141785-10G
Bis(triphenylphosphine)nickel(II) chloride ≥98%
Bis(triphenylphosphine)nickel (II) chloride
- RTECS: QR6170000
- Hygroscopic
- Powder
Formula:
C₃₆H₃₂Cl₂NiP₂ MW: 656,19 g/mol Melting Pt: 250 °C (dec.) |
MDL Number:
MFCD00009592 CAS Number: 14264-16-5 EINECS: 238-154-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...