Phthalazine ≥98%

Supplier: BeanTown Chemical

Synonyms: Benzo[d]pyridazine

141750-5G 141750-25G
BT141750-25GEA 252.39 USD
BT141750-25G BT141750-5G
Phthalazine ≥98%
Phthalazine

  • Crystalline

Formula: C₈H₆N₂
MW: 130.15 g/mol
Boiling Pt: 317 °C
Melting Pt: 89…92 °C
MDL Number: MFCD00006908
CAS Number: 253-52-1
EINECS: 205-963-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR