Methylurea ≥98%

Supplier: BeanTown Chemical

Synonyms: MethylcarbamideN-Methylurea1-Methylurea

141705-1KG 141705-250G
BT141705-1KGEA 205.63 USD
BT141705-1KG BT141705-250G
Methylurea ≥98%
Methylurea

  • RTECS: YT7175000
  • Crystalline

Formula: C₂H₆N₂O
MW: 74.08 g/mol
Melting Pt: 90 °C
MDL Number: MFCD00007950
CAS Number: 598-50-5
EINECS: 209-935-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR