Ferrocenecarbaldehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: Ferrocenecarboxaldehyde

141070-1G 141070-5G
BT141070-1GEA 17.53 USD
BT141070-1G BT141070-5G
Ferrocenecarbaldehyde ≥97%
Ferrocenecarbaldehyde

  • Air Sensitive
  • Crystalline

Formula: C₁₁H₁₀FeO
MW: 214.04 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD00001429
EINECS: 235-158-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR