Isoquinoline-4-amine ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-AminoisoquinolineIsoquinolin-4-amine

140095-1G
BT140095-1GEA 55.64 USD
BT140095-1G
Isoquinoline-4-amine ≥97%
Isoquinoline-4-amine

  • Solid

Formula: C₉H₈N₂
MW: 144.1700 g/mol
Melting Pt: 107…113 °C
MDL Number: MFCD00034752
CAS Number: 23687-25-4
EINECS: 245-823-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR