Molybdic acid ≥85.0% ACS
Supplier: BeanTown Chemical
139840-500G
139840-2KG
BT139840-500GEA
199.23
USD
BT139840-500G
BT139840-2KG
Molybdic acid ≥85.0% ACS
Molybdic acid
- Crystalline/Powder
Formula:
H₂MoO₄ MW: 161.95 g/mol Melting Pt: 300 °C Density: 3.1 g/mL |
MDL Number:
MFCD00036287 CAS Number: 7782-91-4 EINECS: 231-970-5 |
Specification Test Results
Assay, as MO3 | 85.0% min. |
Insoluble in dilute ammonium hydroxide | ≤0.01% |
Chloride | ≤0.002% |
Arsenate, phosphate, and silicate (as SiO2) | ≤0.001% |
Phosphate (PO4) | ≤5ppm |
Sulfate (SO4) | ≤0.2% |
Heavy metals (as Pb) | ≤0.003% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...