1-Naphthaldehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: α-Naphthaldehyde

139665-50G 139665-1KG 139665-250G
BT139665-1KGEA 460.13 USD
BT139665-1KG BT139665-50G BT139665-250G
1-Naphthaldehyde ≥97%
1-Naphthaldehyde

  • RTECS: QJ0190000
  • Refractive index: 1.652
  • Air Sensitive
  • Liquid

Formula: C₁₁H₈O
MW: 156.18 g/mol
Boiling Pt: 160…161 °C/15 mmHg
Melting Pt: 1…2 °C
Density: 1.15 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00004003
CAS Number: 66-77-3
EINECS: 200-633-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR