(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate

Supplier: BeanTown Chemical

Synonyms: (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate(S,S)-Et-DUPHOS-Rh(+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

139530-250MG
BT139530-250MGEA 439.37 USD
BT139530-250MG
(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate
(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate

  • Air Sensitive, Moisture Sensitive
  • Crystalline

Formula: C₃₁H₄₈F₃O₃P₂RhS
MW: 722.63 g/mol
MDL Number: MFCD00269861
CAS Number: 142184-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR