Ammonium hexachlororhodate(III) hydrate ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Rhodium(III)-ammonium chloride hydrate
139345-5G
139345-1G
139345-250MG
BT139345-1GEA
320.95
USD
BT139345-1G
BT139345-5G
BT139345-250MG
Ammonium hexachlororhodate(III) hydrate ≥99.99% (trace metals basis)
Ammonium hexachlororhodate (III) monohydrate
- Hygroscopic
- Powder
Formula:
H₁₂Cl₆N₃Rh·H₂O MW: 369.74 g/mol (anhy) |
MDL Number:
MFCD00003394 CAS Number: 15336-18-2 EINECS: 239-364-2 |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...