Basic fuchsin

Synonyms: Basic-fuchsinPararosanilineFuchsin basicBasic fuchsin hydrochlorideDiamondfuchsinBasic Red 9(4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochlorideBasic fuchsinCI 425104-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochlorideMagenta ORosaniline hydrochloride

139325-5G 139325-25G 139325-100G
BT139325-25GEA 91.61 USD
BT139325-25G BT139325-5G BT139325-100G
Basic fuchsin
Basic Fuchsin

  • RTECS: CX9850000
  • Crystalline/Powder

Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
MDL Number: MFCD00012569
CAS Number: 632-99-5
EINECS: 211-189-6
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR