6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene

Supplier: BeanTown Chemical

Synonyms: 6-Methoxy-2-(4-methoxyphenyl)-1-benzothiophene

139260-25G
BT139260-25GEA 257.57 USD
BT139260-25G
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene

  • Solid

Formula: C₁₆H₁₄O₂S
MW: 270.35 g/mol
Melting Pt: 191…197 °C
MDL Number: MFCD02083138
CAS Number: 63675-74-1
EINECS: 264-408-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR