7-Bromo-1-chloroisoquinoline ≥97%

Supplier: BeanTown Chemical

139060-5G 139060-1G
BT139060-1GEA 67.31 USD
BT139060-1G BT139060-5G
7-Bromo-1-chloroisoquinoline ≥97%
7-Bromo-1-chloroisoquinoline

  • Solid

Formula: C₉H₅BrClN
MW: 242.50 g/mol
Melting Pt: 121…126 °C
MDL Number: MFCD04039320
CAS Number: 215453-51-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR