2-Aminophenylboronic acid pinacol ester ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Aminobenzeneboronic acid pinacol ester2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

139010-1G 139010-5G
BT139010-1GEA 81.53 USD
BT139010-1G BT139010-5G
2-Aminophenylboronic acid pinacol ester ≥97%
2-Aminophenylboronic acid pinacol ester

  • Air Sensitive
  • Solid

Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
Melting Pt: 63…68 °C
MDL Number: MFCD02179448
CAS Number: 191171-55-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR