N-Iodosuccinimide ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Iodopyrrolidine-2,5-dione

138720-25G 138720-5G 138720-100G
BT138720-100GEA 495.46 USD
BT138720-100G BT138720-25G BT138720-5G
N-Iodosuccinimide ≥97%
N-Iodosuccinimide

  • RTECS: WN2817000
  • Light Sensitive, Moisture Sensitive
  • Crystalline

Formula: C₄H₄INO₂
MW: 224.99 g/mol
Melting Pt: 202…206 °C
Density: 2.245 g/mL
MDL Number: MFCD00005512
CAS Number: 516-12-1
EINECS: 208-221-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR