Tantalum phosphide ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
138640-5G
138640-1G
BT138640-5GEA
215
USD
BT138640-5G
BT138640-1G
Tantalum phosphide ≥99.5% (trace metals basis)
Tantalum phosphide
- -100 Mesh Powder
Formula:
PTa |
MDL Number:
MFCD00054076 CAS Number: 12037-63-7 EINECS: 234-860-5 |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...