Dirhodium(II) tetrakis(methyl-2-pyrrolidinone-5(S)-carboxylate)acetonitrile, 2-propanol complex

Supplier: BeanTown Chemical

138415-500MG
BT138415-500MGEA 1078.21 USD
BT138415-500MG
Dirhodium(II) tetrakis(methyl-2-pyrrolidinone-5(S)-carboxylate)acetonitrile, 2-propanol complex
Dirhodium(II) tetrakis(methyl-2-pyrrolidinone-5(S)-carboxylate)acetonitrile, 2-propanol complex

  • Powder

Formula: C₂₄H₃₂N₄O₁₂Rh₂
MW: 774.347 g/mol
MDL Number: MFCD00192123
CAS Number: 132435-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR