Ethylboronic acid ≥98%
Supplier: BeanTown Chemical
138175-1G
138175-5G
138175-25G
BT138175-1GEA
43.84
USD
BT138175-1G
BT138175-25G
BT138175-5G
Ethylboronic acid ≥98%
Ethylboronic acid
- Air Sensitive
- Crystalline
Formula:
C₂H₇BO₂ Melting Pt: 84…88 °C |
MDL Number:
MFCD01074536 CAS Number: 4433-63-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...