Diethyl phenylmalonate ≥98%

Synonyms: Diethyl 2-phenylmalonate

138055-1KG 138055-250G
BT138055-1KGEA 124 USD
BT138055-1KG BT138055-250G
Diethyl phenylmalonate ≥98%
Diethyl phenylmalonate

  • Refractive index: 1.491
  • Liquid

Formula: C₁₃H₁₆O₄
MW: 236.27 g/mol
Boiling Pt: 170…172 °C/14 mmHg
Melting Pt: 16 °C
Density: 1.095 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00009144
CAS Number: 83-13-6
EINECS: 201-456-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR