3,5-Diphenyl-1H-pyrazole ≥98%

Supplier: BeanTown Chemical

137980-5G 137980-25G
BT137980-25GEA 283.27 USD
BT137980-25G BT137980-5G
3,5-Diphenyl-1H-pyrazole ≥98%
3,5-Diphenyl-1H-pyrazole

  • Solid

Formula: C₁₅H₁₂N₂
Melting Pt: 199…203 °C
MDL Number: MFCD00039675
CAS Number: 1145-01-3
EINECS: 214-543-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR