Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride

137860-100G
BT137860-100GEA 51.3 USD
BT137860-100G
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥99%
Girard's reagent T (Carbazoylmethyltrimethylammonium chloride)

  • RTECS: CP3150000
  • Hygroscopic
  • Crystalline

Formula: C₅H₁₄ClN₃O
MW: 167.64 g/mol
Melting Pt: 188…192 °C (dec.)
MDL Number: MFCD00012009
CAS Number: 123-46-6
EINECS: 204-629-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR