3-Bromo-5-methylpyridine ≥97%
Supplier: BeanTown Chemical
137820-5G
137820-1G
BT137820-1GEA
139.15
USD
BT137820-1G
BT137820-5G
3-Bromo-5-methylpyridine ≥97%
3-Bromo-5-methylpyridine
- Refractive index: 1.562
- Liquid
Formula:
C₆H₆BrN MW: 172.02 g/mol Density: 1.4869 g/mL Flash Pt: 90 °C (194 °F) |
MDL Number:
MFCD01646141 CAS Number: 3430-16-8 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...