2-Bromethylamine hydrobromide ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Bromoethylamine hydrobromide2-Bromoethylammonium bromide

137575-50G 137575-250G 137575-1KG
BT137575-50GEA 55.17 USD
BT137575-50G BT137575-250G BT137575-1KG
2-Bromethylamine hydrobromide ≥98%
2-Bromethylamine hydrobromide

  • RTECS: KQ8225000
  • Hygroscopic
  • Crystalline

Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 172…176 °C
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR