2-Bromo-4-formylpyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Bromopyridine-4-carboxaldehyde2-Bromo-4-pyridinecarboxaldehyde

137060-1G 137060-5G
BT137060-5GEA 458.05 USD
BT137060-5G BT137060-1G
2-Bromo-4-formylpyridine ≥97%
2-Bromo-4-formylpyridine

  • Air Sensitive
  • Crystalline/Chunks

Formula: C₆H₄BrNO
MW: 186.01 g/mol
MDL Number: MFCD04039311
CAS Number: 118289-17-1
EINECS: 000-000-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR