(2-Naphthoxy)acetic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Naphthoxyacetic acid
136785-2.5KG
136785-500G
136785-100G
BT136785-2.5KGEA
404.04
USD
BT136785-2.5KG
BT136785-500G
BT136785-100G
(2-Naphthoxy)acetic acid ≥97%
(2-Naphthoxy)acetic acid
- RTECS: AI9659000
- Crystalline
Formula:
C₁₂H₁₀O₃ MW: 202.21 g/mol Melting Pt: 151…154 °C |
MDL Number:
MFCD00004066 CAS Number: 120-23-0 EINECS: 204-380-0 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...