Azomethine H hydrate

Supplier: BeanTown Chemical

Synonyms: 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid hydrate8-Hydroxy-1-[[(2-hydroxyphenyl)methylene]amino]naphthalene-3,6-disulfonic acid hydrate4-Hydroxy-5-[[(2-hydroxyphenyl)methylene]amino]naphthalene-2,7-disulphonic acid hydrateAzomethine-H hydrate8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulfonic acid hydrate8-Hydroxy-1-[[(2-hydroxyphenyl)methylene]amino]naphthalene-3,6-disulphonic acid hydrate4-Hydroxy-5-[[(2-hydroxyphenyl)methylene]amino]naphthalene-2,7-disulfonic acid hydrate

136770-5G 136770-1G
BT136770-1GEA 23.48 USD
BT136770-1G BT136770-5G
Azomethine H hydrate
Azomethine H hydrate

  • Crystalline

Formula: C₁₇H₁₃NO₈S₂·H₂O
MW: 441.44 g/mol
MDL Number: MFCD00066536
CAS Number: 32266-60-7
EINECS: 250-975-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR