2-Bromoterephthalic acid ≥97%

Supplier: BeanTown Chemical

136675-5G 136675-100G 136675-25G
BT136675-100GEA 667.87 USD
BT136675-100G BT136675-5G BT136675-25G
2-Bromoterephthalic acid ≥97%
2-Bromoterephthalic acid

  • Powder

Formula: C₈H₅BrO₄
Melting Pt: 295…297 °C
MDL Number: MFCD00002403
CAS Number: 586-35-6
EINECS: 209-572-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR