Hexylphosphonic acid

Supplier: BeanTown Chemical

Synonyms: n-Hexylphosphonic acidN-hexylphosphonicacidHPA

136310-1G 136310-5G
BT136310-1GEA 68.25 USD
BT136310-1G BT136310-5G
Hexylphosphonic acid
Hexylphosphonic acid

  • Powder

Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
Melting Pt: 97…103 °C
MDL Number: MFCD00013961
CAS Number: 4721-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR