1-Benzyl-5-phenylbarbituric acid ≥98%
Supplier: BeanTown Chemical
136175-25G
136175-5G
BT136175-5GEA
41.45
USD
BT136175-5G
BT136175-25G
1-Benzyl-5-phenylbarbituric acid ≥98%
1-Benzyl-5-phenylbarbituric acid
- Solid
Formula:
C₁₇H₁₄N₂O₃ |
MDL Number:
MFCD00082750 CAS Number: 72846-00-5 EINECS: 276-940-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...