Celite® Hyflo Super-cel®, filter aid
Supplier: BeanTown Chemical
136170-2.5KG
136170-500G
BT136170-500GEA
35.71
USD
BT136170-500G
BT136170-2.5KG
Celite® Hyflo Super-cel®, filter aid
Celite® Hyflo® Super Cel, filter aid
- RTECS: VV7311000
- Powder
MDL Number:
MFCD00132803 CAS Number: 61790-53-2 EINECS: 319-127-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...