2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Methoxy-N,6-dimethyl-1,3,5-triazin-2-amine

136135-25G
BT136135-25GEA 184.89 USD
BT136135-25G
2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine ≥98%
2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine

  • RTECS: XY2905180
  • Solid

Formula: C₆H₁₀N₄O
MW: 154.17 g/mol
Melting Pt: 162…166 °C
MDL Number: MFCD00585858
CAS Number: 5248-39-5
EINECS: 401-360-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR