Poly(methyl methacrylate), M.W. ∼550000

Synonyms: PMMAMethyl Methacrylate PolymerMethacrylic Acid Methyl Ester Polymer

136125-50G 136125-250G 136125-1KG
BT136125-250GEA 55.31 USD
BT136125-250G BT136125-1KG BT136125-50G
Poly(methyl methacrylate), M.W. ∼550000
Poly(methyl methacrylate)

  • RTECS: TR0400000
  • Powder

Formula: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
Melting Pt: >150 °C
Density: 1.17 g/mL
MDL Number: MFCD00134349
CAS Number: 9011-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR