Calcium phosphate tribasic
Supplier: BeanTown Chemical
Synonyms:
tri-Calcium phosphate
, Calcium Hydroxide Phosphate
, HAp
, Hydroxylapatite
, Calcium hydroxyphosphate
, Tribasic calcium phosphate
135475-10KG
135475-500G
135475-2KG
BT135475-500GEA
71.27
USD
BT135475-500G
BT135475-2KG
BT135475-10KG
Calcium phosphate tribasic
Calcium phosphate tribasic
- Powder
Formula:
Ca₅HO₁₃P₃ MW: 502.306 g/mol |
MDL Number:
MFCD00010904 CAS Number: 12167-74-7 EINECS: 235-330-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...