HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)
, HBTU
134635-25G
134635-5G
134635-100G
BT134635-5GEA
63.88
USD
BT134635-5G
BT134635-25G
BT134635-100G
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98%
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
- Light Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₁₁H₁₆F₆N₅OP MW: 379.2 g/mol Melting Pt: 200 °C (dec.) |
MDL Number:
MFCD00075445 CAS Number: 94790-37-1 EINECS: 423-020-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...