Polydimethylsiloxane, hydroxy terminated, M.W. ∼ 4,200

Synonyms: Polysiloxanes, di-Me, hydroxy-terminated

134585-250ML 134585-1L 134585-50ML
BT134585-250MLEA 46.5 USD
BT134585-250ML BT134585-1L BT134585-50ML
Polydimethylsiloxane, hydroxy terminated, M.W. ∼ 4,200
Polydimethylsiloxane, hydroxy terminated

  • Liquid

Formula: [Si(CH₃)₂O]n
MW: 45.092 g/mol
Density: 0.97 g/mL
MDL Number: MFCD01325012
CAS Number: 70131-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR