(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole ≥97%
Supplier: BeanTown Chemical
Synonyms:
(S)-N-Despropyl Pramipexole
134580-1G
BT134580-1GEA
81.53
USD
BT134580-1G
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole ≥97%
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole
- Light Sensitive, Moisture Sensitive
- Solid
Formula:
C₇H₁₁N₃S MW: 169.25 g/mol Melting Pt: 230…231 °C |
MDL Number:
MFCD07368003 CAS Number: 106092-09-5 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...